Learn More
Medchemexpress LLC D-erythro-dihydrosphingosine | 764-22-7 | MFCD07371371 | 98.0% | 301.51 g·mol⁻1 | C18H39NO2 | 10 MG
SDP
Catalog No. 5000299789
Encompass_Preferred
D-erythro-dihydrosphingosine is an endogenous sphingoid base used as a research reagent and biochemical standard. It is a white to off-white solid (CAS 764-22-7) with formula C18H39NO2 and molecular weight 301.51 g·mol⁻1, and functions as a direct inhibitor of cytosolic phospholipase A2α (cPLA2α).
  • Used as a biochemical standard in lipid signaling studies.
  • Directly inhibits cytosolic phospholipase A2α (cPLA2α), useful for enzyme activity assays.
  • Available in multiple small pack sizes suitable for analytical and preparative work.
  • Solid, stable form simplifies storage and handling.
  • Supplied with datasheet, SDS, COA, and LCMS documentation for quality control.
Catalog No. 50-002-99789 Supplier Medchemexpress LLC Supplier No. HYW01983810MG
D-erythro-dihydrosphingosine is an endogenous sphingoid base used as a research reagent and biochemical standard. It is a white to off-white solid (CAS 764-22-7) with formula C18H39NO2 and molecular weight 301.51 g·mol⁻1, and functions as a direct inhibitor of cytosolic phospholipase A2α (cPLA2α).
  • Used as a biochemical standard in lipid signaling studies.
  • Directly inhibits cytosolic phospholipase A2α (cPLA2α), useful for enzyme activity assays.
  • Available in multiple small pack sizes suitable for analytical and preparative work.
  • Solid, stable form simplifies storage and handling.
  • Supplied with datasheet, SDS, COA, and LCMS documentation for quality control.