Learn More
Medchemexpress LLC D-erythro-dihydrosphingosine | 764-22-7 | MFCD07371371 | 98.0% | 301.51 g/mol | C18H39NO2 | 1 ML
SDP
Catalog No. 5000300992
Encompass_Preferred
D-erythro-dihydrosphingosine is an endogenous sphingoid base (sphinganine) and bioactive lipid that directly inhibits cytosolic phospholipase A2α (cPLA2α). Supplied as an analytical standard in solution, it is used in biochemical assays and lipid research to probe sphingolipid metabolism and enzyme activity.
  • Inhibits cytosolic phospholipase A2α activity.
  • Suitable for biochemical assays and lipid research.
  • Analytical standard with reported high purity.
  • Supplied as a DMSO solution for easy dilution and use.
  • Molecular weight 301.51 g/mol and formula C18H39NO2.
Catalog No. 50-003-00992 Supplier Medchemexpress LLC Supplier No. HYW01983810MM/1ML
D-erythro-dihydrosphingosine is an endogenous sphingoid base (sphinganine) and bioactive lipid that directly inhibits cytosolic phospholipase A2α (cPLA2α). Supplied as an analytical standard in solution, it is used in biochemical assays and lipid research to probe sphingolipid metabolism and enzyme activity.
  • Inhibits cytosolic phospholipase A2α activity.
  • Suitable for biochemical assays and lipid research.
  • Analytical standard with reported high purity.
  • Supplied as a DMSO solution for easy dilution and use.
  • Molecular weight 301.51 g/mol and formula C18H39NO2.