Learn More
Medchemexpress LLC D-erythro-dihydrosphingosine-d7 | 1246304-35-7 | >98.0% | 308.55 | C18H32D7NO2 | 500 UG
SDP
Catalog No. 5000300101
Encompass_Preferred
D-erythro-dihydrosphingosine-d7 is the deuterium-labeled analogue of D-erythro-dihydrosphingosine, provided as an isotopically labeled analytical standard for quantitative and mechanistic lipid research. It is suitable as an internal standard or reference material in experiments where deuterium labeling facilitates accurate measurement and tracing of sphingolipid metabolism.
  • Deuterium-labeled (d7) analogue for use as an internal standard.
  • Suitable for quantitative lipidomics and metabolic studies.
  • Reported to inhibit cytosolic phospholipase A2α (cPLA2α), useful in mechanistic assays.
  • Molecular formula C18H32D7NO2 and molecular weight 308.55 for analytical reference.
  • Provided in small reference quantities appropriate for analytical applications.
Catalog No. 50-003-00101 Supplier Medchemexpress LLC Supplier No. HYW019838S500UG
D-erythro-dihydrosphingosine-d7 is the deuterium-labeled analogue of D-erythro-dihydrosphingosine, provided as an isotopically labeled analytical standard for quantitative and mechanistic lipid research. It is suitable as an internal standard or reference material in experiments where deuterium labeling facilitates accurate measurement and tracing of sphingolipid metabolism.
  • Deuterium-labeled (d7) analogue for use as an internal standard.
  • Suitable for quantitative lipidomics and metabolic studies.
  • Reported to inhibit cytosolic phospholipase A2α (cPLA2α), useful in mechanistic assays.
  • Molecular formula C18H32D7NO2 and molecular weight 308.55 for analytical reference.
  • Provided in small reference quantities appropriate for analytical applications.