Learn More
Medchemexpress LLC D-erythro-MAPP (D-e-MAPP) | 143492-38-0 | MFCD01076527 | 98.0% | 361.56 g·mol⁻¹ | C23H39NO2 | 5 MG
SDP
Catalog No. 5000223510
Encompass_Preferred
D-erythro-MAPP is a research reagent that inhibits ceramidase activity and is used to study ceramide metabolism, cell viability, and cell-cycle regulation. Reported in vitro potency is in the low micromolar range, and cellular studies show reduced viability and G0/G1 arrest in some cancer cell lines.
  • Inhibits ceramidase with low micromolar potency
  • Reduces cell viability and induces G0/G1 cell-cycle arrest in cell assays
  • Solid, white to off-white appearance with 98.0% purity
  • Soluble in DMF and ethanol at ≥20 mg/mL
  • Stable as powder at -20°C for up to 3 years
  • Available in small pack sizes suitable for laboratory research
Catalog No. 50-002-23510 Supplier Medchemexpress LLC Supplier No. HY1374225MG
D-erythro-MAPP is a research reagent that inhibits ceramidase activity and is used to study ceramide metabolism, cell viability, and cell-cycle regulation. Reported in vitro potency is in the low micromolar range, and cellular studies show reduced viability and G0/G1 arrest in some cancer cell lines.
  • Inhibits ceramidase with low micromolar potency
  • Reduces cell viability and induces G0/G1 cell-cycle arrest in cell assays
  • Solid, white to off-white appearance with 98.0% purity
  • Soluble in DMF and ethanol at ≥20 mg/mL
  • Stable as powder at -20°C for up to 3 years
  • Available in small pack sizes suitable for laboratory research