Learn More
Medchemexpress LLC D-erythro-neopterin-13C5 | 1217632-04-6 | 97.5% | 258.18 g·mol⁻¹ | C9H11N5O4 (13C5) | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW040055S1MG
Encompass_Preferred
Neopterin-13C5 is a 13C5 isotopically labeled form of D-erythro-neopterin supplied for research use as an internal standard or tracer in quantitative mass spectrometry and metabolic studies. It is provided as a small, solid quantity suitable for analytical workflows.
  • Isotopically labeled (13C5) internal standard for mass spectrometry.
  • Suitable as a tracer in metabolic and pharmacokinetic studies.
  • Reported high purity (~97.5%).
  • Molecular weight 258.18 g·mol⁻¹ for accurate mass calibration.
  • Supplied in small milligram quantities for analytical assays.
  • For research use only; not for human or therapeutic use.
Catalog No. 50-002-99501