Learn More
Abcam D-erythro Sphingosine-1-phosphate, agonist of S1P receptors, 1MG
Catalog No. 502706447
Encompass_Preferred
MW 379.5 Da, Purity >98%. Endogenous signaling sphingolipid and agonist of S1P receptors (EDG-GPCRs). Mobilises Ca2+ from intracellular stores via IP3 receptor independent pathways. Activates MAPK pathways. Regulates a variety of cellular responses and promotes angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6447 Supplier Abcam Supplier No. AB1417481MG
MW 379.5 Da, Purity >98%. Endogenous signaling sphingolipid and agonist of S1P receptors (EDG-GPCRs). Mobilises Ca2+ from intracellular stores via IP3 receptor independent pathways. Activates MAPK pathways. Regulates a variety of cellular responses and promotes angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement