Learn More
Medchemexpress LLC HY-101047 10mg Medchemexpress, D-erythro-Sphingosine CAS:123-78-4 Purity:>98%
SDP
Catalog No. 501932020
Encompass_Preferred
Medchemexpress, HY-101047 10mg D-erythro-Sphingosine CAS:123-78-4 D-erythro-Sphingosine is a very potent activator of p32-kinase with an EC 50 of 8 μM. D-erythro-Sphingosine inhibits protein kinase C ( PKC ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2020 Supplier Medchemexpress LLC Supplier No. HY10104710MG
Medchemexpress, HY-101047 10mg D-erythro-Sphingosine CAS:123-78-4 D-erythro-Sphingosine is a very potent activator of p32-kinase with an EC 50 of 8 μM. D-erythro-Sphingosine inhibits protein kinase C ( PKC ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.