Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-erythro-sphingosine | 123-78-4 | 299.49 g/mol | C18H37NO2 | 25 MG
SDP

Catalog No. 5000212116
Encompass_Preferred

D-erythro-sphingosine is a research-grade bioactive sphingolipid used to study lipid signaling and kinase regulation. It functions as an activator of p32-kinase (EC50 ≈ 8 μM), an inhibitor of protein kinase C, and a PP2A activator. The material is provided as a powder with manufacturer guidance for storage, solubility, and formulation for in vitro and in vivo use.

  • Activates p32-kinase (EC50 ≈ 8 μM).
  • Inhibits protein kinase C.
  • Activates PP2A and acts as a human endogenous metabolite.
  • Supplied as a powder; store at -20°C for long-term stability.
  • Soluble in DMSO (50 mg/mL) with heating/sonication; in vivo formulation guidance provided.
  • Molecular formula C18H37NO2; molecular weight 299.49 g/mol.

Catalog No. 50-002-12116 Supplier Medchemexpress LLC Supplier No. HY10104725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-erythro-sphingosine is a research-grade bioactive sphingolipid used to study lipid signaling and kinase regulation. It functions as an activator of p32-kinase (EC50 ≈ 8 μM), an inhibitor of protein kinase C, and a PP2A activator. The material is provided as a powder with manufacturer guidance for storage, solubility, and formulation for in vitro and in vivo use.

  • Activates p32-kinase (EC50 ≈ 8 μM).
  • Inhibits protein kinase C.
  • Activates PP2A and acts as a human endogenous metabolite.
  • Supplied as a powder; store at -20°C for long-term stability.
  • Soluble in DMSO (50 mg/mL) with heating/sonication; in vivo formulation guidance provided.
  • Molecular formula C18H37NO2; molecular weight 299.49 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.