Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-erythro-sphingosine | 123-78-4 | MFCD00036751 | >97.0% | 299.49 g/mol | C18H37NO2 | 1 ML
SDP

Catalog No. 5000213623
Encompass_Preferred

D-erythro-sphingosine is a bioactive sphingoid base used in biochemical and cell-signaling research. It functions as a potent activator of p32-kinase (EC50 ≈ 8 μM), an inhibitor of protein kinase C, and an activator of PP2A. The compound is supplied as a ready-to-use solution and as solids for reconstitution for research use only.

  • Acts as a p32-kinase activator (EC50 ≈ 8 μM) and modulates PKC activity.
  • Also activates protein phosphatase 2A, supporting signaling studies.
  • Available as a 10 mM solution in DMSO (1 mL) or as solid for reconstitution.
  • Molecular formula C18H37NO2 and molecular weight 299.49 g/mol.
  • Powder stable at -20°C; in solvent store at -80°C for long-term storage.
  • Intended for research use only and not for human or diagnostic applications.

Catalog No. 50-002-13623 Supplier Medchemexpress LLC Supplier No. HY10104710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-erythro-sphingosine is a bioactive sphingoid base used in biochemical and cell-signaling research. It functions as a potent activator of p32-kinase (EC50 ≈ 8 μM), an inhibitor of protein kinase C, and an activator of PP2A. The compound is supplied as a ready-to-use solution and as solids for reconstitution for research use only.

  • Acts as a p32-kinase activator (EC50 ≈ 8 μM) and modulates PKC activity.
  • Also activates protein phosphatase 2A, supporting signaling studies.
  • Available as a 10 mM solution in DMSO (1 mL) or as solid for reconstitution.
  • Molecular formula C18H37NO2 and molecular weight 299.49 g/mol.
  • Powder stable at -20°C; in solvent store at -80°C for long-term storage.
  • Intended for research use only and not for human or diagnostic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.