Learn More
Medchemexpress LLC D-erythro-sphingosine | 123-78-4 | MFCD00036751 | 98.0% | 299.49 g/mol | C18H37NO2 | 50 MG
SDP
Catalog No. 5000212266
Encompass_Preferred
D-erythro-sphingosine is a research-grade sphingolipid reagent used as a biochemical probe in cell signaling and kinase research. It activates p32-kinase (EC50 ≈ 8 μM), inhibits protein kinase C, and activates PP2A.
  • High purity (98.0%)
  • Molecular weight 299.49 g/mol, formula C18H37NO2
  • Solid powder format for solution preparation
  • Available in multiple sizes, including 50 mg
  • Useful for studies of kinase modulation and sphingolipid pathways
Catalog No. 50-002-12266 Supplier Medchemexpress LLC Supplier No. HY10104750MG
D-erythro-sphingosine is a research-grade sphingolipid reagent used as a biochemical probe in cell signaling and kinase research. It activates p32-kinase (EC50 ≈ 8 μM), inhibits protein kinase C, and activates PP2A.
  • High purity (98.0%)
  • Molecular weight 299.49 g/mol, formula C18H37NO2
  • Solid powder format for solution preparation
  • Available in multiple sizes, including 50 mg
  • Useful for studies of kinase modulation and sphingolipid pathways