Learn More
Enzo Life Sciences D-erythro-Sphingosine (synthetic) (25mg). CAS: 123-78-4
SDP
Catalog No. 502008660
Encompass_Preferred
Common component of cell membranes, often as phosphatidyl conjugates. Inhibitor of protein kinase C (PKC) and calmodulin-dependent enzymes. Purity: ≥97% (TLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8660 Supplier Enzo Life Sciences Supplier No. ALX306011M025
Common component of cell membranes, often as phosphatidyl conjugates. Inhibitor of protein kinase C (PKC) and calmodulin-dependent enzymes. Purity: ≥97% (TLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.