Learn More
TARGETMOL CHEMICALS INC D-ERYTHRO-SPHINGOSINE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T589150MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. D-erythro-Sphingosine (trans-4-Sphingenine) is a protein kinase C (PKC) inhibitor. D-erythro-Sphingosine (Erythrosphingosine) is also a PP2A activator Purity 98%
Catalog No. 50-288-0121