Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-erythro-sphingosine (C20 sphingosine) | 6918-49-6 | MFCD08703032 | 99.0% | 327.55 g·mol⁻¹ | C20H41NO2 | 5 MG
SDP

Catalog No. 5000263094
Encompass_Preferred

C20-Sphingosine is a long-chain sphingoid base (D-erythro-sphingosine, C20) used in studies of neural development and sphingolipid biology. Supplied as a high-purity solid for in vitro research, it is suitable for experiments investigating lipid signaling, metabolism, and cellular development.

  • Long-chain sphingoid base used to study neural development and sphingolipid biology.
  • High purity (≥99.0%).
  • Molecular formula C20H41NO2; molecular weight 327.55 g·mol⁻¹.
  • Supplied as a solid, suitable for dissolution in common organic solvents.
  • Recommended storage: powder at -20°C; in solvent store at -80°C (short term) or -20°C.
  • Intended for research use only; not for clinical or therapeutic use.

Catalog No. 50-002-63094 Supplier Medchemexpress LLC Supplier No. HY1341045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C20-Sphingosine is a long-chain sphingoid base (D-erythro-sphingosine, C20) used in studies of neural development and sphingolipid biology. Supplied as a high-purity solid for in vitro research, it is suitable for experiments investigating lipid signaling, metabolism, and cellular development.

  • Long-chain sphingoid base used to study neural development and sphingolipid biology.
  • High purity (≥99.0%).
  • Molecular formula C20H41NO2; molecular weight 327.55 g·mol⁻¹.
  • Supplied as a solid, suitable for dissolution in common organic solvents.
  • Recommended storage: powder at -20°C; in solvent store at -80°C (short term) or -20°C.
  • Intended for research use only; not for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.