Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-erythro-sphingosine-d7 | 1246304-34-6 | 99.0% | 306.54 | C18H30D7NO2 | 500 UG
SDP

Catalog No. 5000212264
Encompass_Preferred

D-erythro-sphingosine-d7 is the deuterium-labeled analogue of D-erythro-sphingosine supplied for research use. It acts as a p32-kinase activator (EC50 ≈ 8 μM), inhibits protein kinase C, and is used as an isotopic tracer in biochemical and analytical studies.

  • Deuterium-labeled (d7) isotopologue for tracer and quantitation studies.
  • High purity, typically 99.0%.
  • Molecular weight 306.54 g/mol.
  • Chemical formula C18H30D7NO2.
  • Suitable for biochemical assays and mass spectrometry applications.
  • Provided as a white to off-white solid for laboratory use.

Catalog No. 50-002-12264 Supplier Medchemexpress LLC Supplier No. HY101047S500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-erythro-sphingosine-d7 is the deuterium-labeled analogue of D-erythro-sphingosine supplied for research use. It acts as a p32-kinase activator (EC50 ≈ 8 μM), inhibits protein kinase C, and is used as an isotopic tracer in biochemical and analytical studies.

  • Deuterium-labeled (d7) isotopologue for tracer and quantitation studies.
  • High purity, typically 99.0%.
  • Molecular weight 306.54 g/mol.
  • Chemical formula C18H30D7NO2.
  • Suitable for biochemical assays and mass spectrometry applications.
  • Provided as a white to off-white solid for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.