Learn More
Medchemexpress LLC D-erythro-sphingosine (erythrosphingosine) | 123-78-4 | MFCD00036751 | 98.0% | 299.49 g/mol | C18H37NO2 | 5 MG
SDP
Catalog No. 5000212117
Encompass_Preferred
D-erythro-Sphingosine is a sphingoid base used as a biochemical reagent and signaling lipid. It modulates sphingolipid pathways and acts as a protein kinase C inhibitor, making it useful for cell signaling research and enzyme inhibition assays. The compound is typically supplied as a high-purity solid for laboratory use.
  • Acts as a protein kinase C inhibitor.
  • Used in cell signaling and enzyme inhibition assays.
  • High purity suitable for analytical and biological research.
  • Molecular formula C18H37NO2 and molecular weight 299.49 g/mol.
  • Available in small laboratory quantities for experimental use.
Catalog No. 50-002-12117 Supplier Medchemexpress LLC Supplier No. HY1010475MG
D-erythro-Sphingosine is a sphingoid base used as a biochemical reagent and signaling lipid. It modulates sphingolipid pathways and acts as a protein kinase C inhibitor, making it useful for cell signaling research and enzyme inhibition assays. The compound is typically supplied as a high-purity solid for laboratory use.
  • Acts as a protein kinase C inhibitor.
  • Used in cell signaling and enzyme inhibition assays.
  • High purity suitable for analytical and biological research.
  • Molecular formula C18H37NO2 and molecular weight 299.49 g/mol.
  • Available in small laboratory quantities for experimental use.