Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-erythro-sphingosine (erythrosphingosine) | 123-78-4 | MFCD00036751 | 98.0% | 299.49 g/mol | C18H37NO2 | 500 MG
SDP

Catalog No. 5000212060
Encompass_Preferred

D-erythro-Sphingosine is a research-grade sphingoid base used as a biochemical tool compound. It functions as a potent activator of p32-kinase and an inhibitor of protein kinase C, and is supplied as a purified analytical standard for in vitro and in vivo studies.

  • High purity: 98.0%.
  • Molecular formula C18H37NO2 and molecular weight 299.49 g/mol.
  • Available in multiple quantities, including 500 mg.
  • Powder storage at -20°C is stable for up to 3 years.
  • Soluble in DMSO at 50 mg/mL; aliquot solutions to avoid repeated freeze-thaw cycles.
  • In vivo formulation protocols available for dosing studies.

Catalog No. 50-002-12060 Supplier Medchemexpress LLC Supplier No. HY101047500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-erythro-Sphingosine is a research-grade sphingoid base used as a biochemical tool compound. It functions as a potent activator of p32-kinase and an inhibitor of protein kinase C, and is supplied as a purified analytical standard for in vitro and in vivo studies.

  • High purity: 98.0%.
  • Molecular formula C18H37NO2 and molecular weight 299.49 g/mol.
  • Available in multiple quantities, including 500 mg.
  • Powder storage at -20°C is stable for up to 3 years.
  • Soluble in DMSO at 50 mg/mL; aliquot solutions to avoid repeated freeze-thaw cycles.
  • In vivo formulation protocols available for dosing studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.