Learn More
Medchemexpress LLC D-erythro-sphingosine hydrochloride | 2673-72-5 | 99.9% | 335.95 | C18H38ClNO2 | 25 MG
SDP
Catalog No. 5000208646
Encompass_Preferred
D-erythro-sphingosine hydrochloride is a small-molecule sphingolipid used in research to activate TRPM3 ion channels and modulate protein phosphorylation pathways, including retinoblastoma protein dephosphorylation. It is provided as a high-purity solid for cellular signaling and kinase modulation studies.
  • Specific TRPM3 activator.
  • Modulates retinoblastoma protein phosphorylation.
  • High purity (99.9%) solid powder.
  • Suitable for cellular signaling and kinase assays.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.
Catalog No. 50-002-08646 Supplier Medchemexpress LLC Supplier No. 25MGHYW089922
D-erythro-sphingosine hydrochloride is a small-molecule sphingolipid used in research to activate TRPM3 ion channels and modulate protein phosphorylation pathways, including retinoblastoma protein dephosphorylation. It is provided as a high-purity solid for cellular signaling and kinase modulation studies.
  • Specific TRPM3 activator.
  • Modulates retinoblastoma protein phosphorylation.
  • High purity (99.9%) solid powder.
  • Suitable for cellular signaling and kinase assays.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.