Learn More
Medchemexpress LLC D-Erythrose-4-13C 10mg | 90913-08-9 | 10 MG
SDP
Catalog No. 5000226598
Encompass_Preferred
D-Erythrose-4-13C is the 13C-labeled isotopologue of D-erythrose supplied as a small-volume solution for use as a stable isotope tracer or analytical standard in metabolic and biochemical studies.
  • 13C label at the 4-carbon position, indicated by the product SMILES.
  • High chemical purity of 99.18%.
  • Supplied as a colorless to light yellow liquid solution.
  • Available in small pre-weighed sizes suitable for analytical workflows.
  • Intended for tracer studies, NMR, mass spectrometry, and analytical reference standards.
Catalog No. 50-002-26598 Supplier Medchemexpress LLC Supplier No. HY116956S310MG
D-Erythrose-4-13C is the 13C-labeled isotopologue of D-erythrose supplied as a small-volume solution for use as a stable isotope tracer or analytical standard in metabolic and biochemical studies.
  • 13C label at the 4-carbon position, indicated by the product SMILES.
  • High chemical purity of 99.18%.
  • Supplied as a colorless to light yellow liquid solution.
  • Available in small pre-weighed sizes suitable for analytical workflows.
  • Intended for tracer studies, NMR, mass spectrometry, and analytical reference standards.