Learn More
Medchemexpress LLC D-Erythrose-4-13C 25mg | 90913-08-9 | 121.10 g/mol | C4H8O4 (13C-labeled at C-4) | 25 MG
SDP
Catalog No. 5000227904
Encompass_Preferred
D-Erythrose-4-13C is the carbon-13 labeled form of D-erythrose supplied as an aqueous solution for laboratory research. It is used as a stable-isotope tracer and as an internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. Typical purity is 99.18% and the material is stored at -20°C for long-term stability.
  • Used as a stable-isotope tracer for metabolic and pathway studies.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Provided as an aqueous solution for convenient handling.
  • High chemical purity for accurate analytical measurements.
  • Store at -20°C for long-term stability.
Catalog No. 50-002-27904 Supplier Medchemexpress LLC Supplier No. HY116956S325MG
D-Erythrose-4-13C is the carbon-13 labeled form of D-erythrose supplied as an aqueous solution for laboratory research. It is used as a stable-isotope tracer and as an internal standard for quantitative analyses by NMR, GC-MS, and LC-MS. Typical purity is 99.18% and the material is stored at -20°C for long-term stability.
  • Used as a stable-isotope tracer for metabolic and pathway studies.
  • Suitable as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Provided as an aqueous solution for convenient handling.
  • High chemical purity for accurate analytical measurements.
  • Store at -20°C for long-term stability.