Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(-)-Erythrose 85%, Thermo Scientific™

Catalog No. AC153122500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153122500 250 mg
AC153120010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153122500 Supplier Thermo Scientific Chemicals Supplier No. 153122500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-50-6
Molecular Formula C4H8O4
Molecular Weight (g/mol) 120.104
InChI Key YTBSYETUWUMLBZ-IUYQGCFVSA-N
Synonym d-erythrose, erythrose, d---erythrose, 2r,3r-2,3,4-trihydroxybutanal, d-erythro-tetrose, unii-x3ei0we8q4, r*,r*-2,3,4-trihydroxybutanal, x3ei0we8q4, butanal, 2,3,4-trihydroxy-, r*,r*, erythrose, d
PubChem CID 94176
ChEBI CHEBI:27904
IUPAC Name (2R,3R)-2,3,4-trihydroxybutanal
SMILES C(C(C(C=O)O)O)O

Specifications

Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4990 to 1.5010 (20°C, 589nm)
Linear Formula HOCH2[CH(OH)]2CHO
Quantity 250 mg
Beilstein 31, 12
Merck Index 15, 3746
Specific Rotation After 6 day: -29° to -31° (20°C, 589nm) (c=2, H2O/NH3)
Formula Weight 120.1
Percent Purity ≥80%
Physical Form Very Viscous Liquid
Chemical Name or Material D(-)-Erythrose, 85%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 209-505-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.