Learn More
TARGETMOL CHEMICALS INC D-F07 1MG
SDP
Catalog No. 502879754
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. D-F07 is a novel fluorescent IRE-1 RNase inhibitor and a tricyclic chromone with potential anticancer activity. Purity 99.47%
Catalog No. 50-287-9754 Supplier TARGETMOL CHEMICALS INC Supplier No. T843111MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. D-F07 is a novel fluorescent IRE-1 RNase inhibitor and a tricyclic chromone with potential anticancer activity. Purity 99.47%