Learn More
Chondrex Inc Dust Mite Extract from D. fainae, 0.6 mg, lyophilzed
Supplier:  Chondrex Inc 3040
Encompass_Preferred
House dust mite (HDM) is the most important indoor allergen for humans. Two main mite species, Dermatophagoides pteronyssinus (D. pteronyssinus) and Dermatophagoides farinae (D. farinae) are the major allergens responsible for asthma in human allergy patients. This HDM can be used for inducing HDM-induced asthma mouse models.
Catalog No. 50-165-6746