Learn More
Leica Biosystems D FORMALIZER PAD/LARGE ROLL
Catalog No. NC0320979
Encompass_Preferred
D-Formalizer Pads reduce formaldehyde exposure by absorbing and neutralizing any excess formalin. The durable blue surface of the pads provides an excellent contrast to even small pieces of tissue. The Pads are also useful for soaking up leakage and de-odourising vapours in other applications. They can be used to line shelves which hold specimen containers that may seep formaldehyde around the lid, or to line transport boxes where leaks may occur. D-Formalizer Pads are available in various sizes and also in a convenient roll so that lengths can be cut as required.
Catalog No. NC0320979 Supplier Leica Biosystems Supplier No. 380075314600
D-Formalizer Pads reduce formaldehyde exposure by absorbing and neutralizing any excess formalin. The durable blue surface of the pads provides an excellent contrast to even small pieces of tissue. The Pads are also useful for soaking up leakage and de-odourising vapours in other applications. They can be used to line shelves which hold specimen containers that may seep formaldehyde around the lid, or to line transport boxes where leaks may occur. D-Formalizer Pads are available in various sizes and also in a convenient roll so that lengths can be cut as required.