Learn More
Cayman Chemical D-Fructose-1,6-bisphosphate (sodium salt hydrate), Purity: ≥95%, CAS: 81028-91-3, 1 g
SDP
Catalog No. 502984483
Encompass_Preferred
An intermediate in carbohydrate metabolism, including glycolysis and gluconeogenesis
Catalog No. 50-298-4483 Supplier Cayman Chemical Supplier No. 205161 G
An intermediate in carbohydrate metabolism, including glycolysis and gluconeogenesis