Learn More
Cayman Chemical D-Fructose-1,6-bisphosphate (sodium salt hydrate), Purity: ≥95%, CAS: 81028-91-3, 5 g
SDP
Catalog No. 502984363
Encompass_Preferred
An intermediate in carbohydrate metabolism, including glycolysis and gluconeogenesis
Catalog No. 50-298-4363 Supplier Cayman Chemical Supplier No. 205165 G
An intermediate in carbohydrate metabolism, including glycolysis and gluconeogenesis