Learn More
AdipoGen D-Fructose 1,6-diphosphate trisodium salt hydrate,
SDP
Supplier:  AdipoGen CDXF0218G001
Encompass_Preferred
D-Fructose 1,6-diphosphate trisodium salt hydrate, AdipoGen Life Sciences. Chemical. CAS: 38099-82-0. Formula: C6H11Na3O12P2 . xH2O. MW: 406.06 (anhydrous basis). Synthetic. D-Fructose-1,6-bisphosphate (FBP), a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolase(s) and fructose-1, 6-bisphosphatase(s). FBP is studied as a neuroprotective agent in brain injury.
Catalog No. 50-596-210