Learn More
AdipoGen D-Fructose 1 6-diphosphate 3Na
SDP
Catalog No. 502560861
Encompass_Preferred
Chemical. CAS 38099-82-0. Formula C6H11Na3O12P2 . xH2O. MW 406.06 anhydrous basis. Synthetic. D-Fructose-1,6-bisphosphate FBP, a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolases and fructose-1, 6-bisphosphatases. FBP is studied as a neuroprotective agent in brain injury.
Catalog No. 50-256-0861 Supplier AdipoGen Supplier No. CDXF0218G005
Chemical. CAS 38099-82-0. Formula C6H11Na3O12P2 . xH2O. MW 406.06 anhydrous basis. Synthetic. D-Fructose-1,6-bisphosphate FBP, a common metabolic sugar, is the precursor of glyceraldehyde 3-phosphate and dihydroxyacetone phosphate in the glycolytic pathway. It is an allosteric activator of enzymes such as pyruvate kinase and a substrate used to identify and characterize enzymes such as fructose-1,6-bisphosphate aldolases and fructose-1, 6-bisphosphatases. FBP is studied as a neuroprotective agent in brain injury.