Learn More
Medchemexpress LLC D-Fructose-13C-1 | 117013-19-1 | 100 MG
SDP
Catalog No. 5000419874
Encompass_Preferred
D-Fructose-13C-1 is the 13C labeled D-Fructose. D-Fructose (D(-)-Fructose) is a naturally occurring monosaccharide found in many plants. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity of 98.0%
  • Appears as a solid, white to off-white
  • Stable isotope
Catalog No. 50-004-19874 Supplier Medchemexpress LLC Supplier No. HYN7092S2100MG
D-Fructose-13C-1 is the 13C labeled D-Fructose. D-Fructose (D(-)-Fructose) is a naturally occurring monosaccharide found in many plants. It is for research use only and not sold to patients.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity of 98.0%
  • Appears as a solid, white to off-white
  • Stable isotope