Learn More
Medchemexpress LLC D-Fructose-13C-1 | 117013-19-1 | 25 MG
SDP
Catalog No. 5000420290
Encompass_Preferred
D-Fructose-13C-1 is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. It is intended for research use only.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope of carbon-13.
Catalog No. 50-004-20290 Supplier Medchemexpress LLC Supplier No. 25MGHYN7092S2
D-Fructose-13C-1 is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. It is intended for research use only.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotope of carbon-13.