Learn More
Medchemexpress LLC D-Fructose-13C-1 | 117013-19-1 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S25MG
Encompass_Preferred
D-Fructose-13C-1 is a 13C labeled D-Fructose, a naturally occurring monosaccharide found in plants. This compound is intended for research use only and can be utilized as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules primarily as tracers for quantitation during drug development, impacting pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-21090