Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose-13C | 108311-21-3 | 25 MG
SDP

Catalog No. 5000421870
Encompass_Preferred

D-Fructose-13C is a 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. It is intended for research use only, serving as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.

  • Purity: 99.95%
  • Appearance: Solid
  • Color: White to off-white
  • Stable heavy isotopes can be incorporated into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-004-21870 Supplier Medchemexpress LLC Supplier No. 25MGHYN7092S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose-13C is a 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. It is intended for research use only, serving as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS.

  • Purity: 99.95%
  • Appearance: Solid
  • Color: White to off-white
  • Stable heavy isotopes can be incorporated into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.