Learn More
Medchemexpress LLC D-Fructose-13C | 108311-21-3 | 5 MG
SDP
Catalog No. 5000418693
Encompass_Preferred
D-Fructose-13C is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated into drug molecules primarily as tracers for quantitation during drug development, with deuteration specifically noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • 13C labeled D-fructose, a naturally occurring monosaccharide
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-18693 Supplier Medchemexpress LLC Supplier No. HYN7092S15MG
Add to Cart
Add to Cart
D-Fructose-13C is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Stable heavy isotopes are incorporated into drug molecules primarily as tracers for quantitation during drug development, with deuteration specifically noted for its potential impact on pharmacokinetic and metabolic profiles of drugs.
  • 13C labeled D-fructose, a naturally occurring monosaccharide
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs