Learn More
Medchemexpress LLC D-Fructose-13C | 108311-21-3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S150MG
Encompass_Preferred
D-Fructose-13C is the 13C labeled D-Fructose. D-Fructose (D(-)-Fructose) is a naturally occurring monosaccharide found in many plants. This product is for research use only and is not sold to patients. The incorporation of stable heavy isotopes like hydrogen, carbon, and other elements into drug molecules serves primarily for quantitation as tracers during drug development. Deuteration, specifically, has gained recognition for its potential influence on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-19890