Learn More
Medchemexpress LLC D-Fructose-13C2 | 2483736-14-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN7092S31MG
Encompass_Preferred
D-Fructose-13C2 is a 13C labeled D-Fructose, which is a naturally occurring monosaccharide found in many plants. This compound is intended for research use only, serving as a tracer and an internal standard for quantitative analysis in various analytical methods.
  • Used as a tracer in research applications
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for quantitation during drug development processes
Catalog No. 50-004-21091