Learn More
Medchemexpress LLC D-Fructose-13C2 | 2483736-14-5 | 10 MG
SDP
Catalog No. 5000419745
Encompass_Preferred
D-Fructose-13C2 is the 13C labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. This compound serves as a valuable tool for research, particularly as a tracer and an internal standard in various analytical techniques.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-19745 Supplier Medchemexpress LLC Supplier No. HYN7092S310MG
D-Fructose-13C2 is the 13C labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. This compound serves as a valuable tool for research, particularly as a tracer and an internal standard in various analytical techniques.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS