Learn More
Medchemexpress LLC D-Fructose-13C2 | 2483736-14-5 | 25 MG
SDP
Catalog No. 5000421798
Encompass_Preferred
D-Fructose-13C2 is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-21798 Supplier Medchemexpress LLC Supplier No. 25MGHYN7092S3
D-Fructose-13C2 is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS