Learn More
Medchemexpress LLC D-Fructose-13C2 | 2483736-14-5 | 5 MG
SDP
Catalog No. 5000421089
Encompass_Preferred
D-Fructose-13C2 is the 13C labeled D-Fructose. D-Fructose (D(-)-Fructose) is a naturally occurring monosaccharide found in many plants. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-21089 Supplier Medchemexpress LLC Supplier No. HYN7092S35MG
D-Fructose-13C2 is the 13C labeled D-Fructose. D-Fructose (D(-)-Fructose) is a naturally occurring monosaccharide found in many plants. This product is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.