Learn More
Medchemexpress LLC D-Fructose-13C2 | 2483736-14-5 | 50 MG
SDP
Catalog No. 5000418262
Encompass_Preferred
D-Fructose-13C2 is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. Intended for research use only, this compound serves as a tracer and an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-18262 Supplier Medchemexpress LLC Supplier No. HYN7092S350MG
D-Fructose-13C2 is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. Intended for research use only, this compound serves as a tracer and an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS