Learn More
Cayman Chemical D-Fructose-13C6 250mg
SDP
Catalog No. 502988612
Encompass_Preferred
An internal standard for the quantification of D-fructose by GC- or LC-MS
Catalog No. 50-298-8612 Supplier Cayman Chemical Supplier No. 30071250 MG
An internal standard for the quantification of D-fructose by GC- or LC-MS