Learn More
Bioworld D-Fructose, 1 kg
Catalog No. 50255982
Encompass_Preferred
A monosaccharide and isomer of glucose Specifications:High Purity GradeMoisture: <0.5%Chloride: <0.018%Sulfate: <0.025%Calcium and Magnesium: <0.005%Heavy metals (as Pb): <0.0005%HS Code : 17025000IUPAC_Name: FructoseMelting Point: 103 °CRTECS Number: LS7120000
Catalog No. 50-255-982 Supplier Bioworld Supplier No. 406000081
A monosaccharide and isomer of glucose Specifications:High Purity GradeMoisture: <0.5%Chloride: <0.018%Sulfate: <0.025%Calcium and Magnesium: <0.005%Heavy metals (as Pb): <0.0005%HS Code : 17025000IUPAC_Name: FructoseMelting Point: 103 °CRTECS Number: LS7120000