Learn More
Medchemexpress LLC D-Fructose-4-13C | 84270-09-7 | 1 MG

Supplier: Medchemexpress LLC HYN7092S51MG
D-Fructose-4-13C is the 13C labeled D-Fructose. D-Fructose is a naturally occurring monosaccharide found in many plants. This product is for research use only.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
- Purity: 99.7%
- Appearance: Solid
- Color: Off-white to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.