Learn More
Medchemexpress LLC D-Fructose-4-13C | 84270-09-7 | 10 MG
SDP
Catalog No. 5000419968
Encompass_Preferred
D-Fructose-4-13C is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. This compound is intended for research use only.
  • Used as a tracer for quantitative analysis
  • Used as an internal standard for quantitative analysis
  • Suitable for NMR analysis
  • Suitable for GC-MS analysis
  • Suitable for LC-MS analysis
Catalog No. 50-004-19968 Supplier Medchemexpress LLC Supplier No. HYN7092S510MG
Add to Cart
Add to Cart
D-Fructose-4-13C is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants. This compound is intended for research use only.
  • Used as a tracer for quantitative analysis
  • Used as an internal standard for quantitative analysis
  • Suitable for NMR analysis
  • Suitable for GC-MS analysis
  • Suitable for LC-MS analysis