Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose-4-13C | 84270-09-7 | 25 MG
SDP

Catalog No. 5000421868
Encompass_Preferred

D-Fructose-4-13C is a 13C labeled D-Fructose, a naturally occurring monosaccharide found in various plants. This compound is suitable for research applications.

  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 99.7%
  • Molecular weight: 181.15
  • Formula: C₅¹³CH₁₂O₆
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage for powder: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-004-21868 Supplier Medchemexpress LLC Supplier No. 25MGHYN7092S5
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose-4-13C is a 13C labeled D-Fructose, a naturally occurring monosaccharide found in various plants. This compound is suitable for research applications.

  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 99.7%
  • Molecular weight: 181.15
  • Formula: C₅¹³CH₁₂O₆
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Storage for powder: -20°C for 3 years
  • Storage in solvent: -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.