Learn More
Medchemexpress LLC D-Fructose-4-13C | 84270-09-7 | 5 MG
SDP
Catalog No. 5000418321
Encompass_Preferred
D-Fructose-4-13C is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-18321 Supplier Medchemexpress LLC Supplier No. HYN7092S55MG
Add to Cart
Add to Cart
D-Fructose-4-13C is the 13C labeled D-Fructose, a naturally occurring monosaccharide found in many plants.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS