Learn More
Medchemexpress LLC D-Fructose-4-13C | 84270-09-7 | 50 MG
SDP
Catalog No. 5000417649
Encompass_Preferred
D-Fructose-4-13C is a 13C labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. It is a solid with an off-white to light yellow appearance and is suitable for research applications. Stable heavy isotopes, such as those in this product, are used as tracers for quantitation during drug development processes.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Supports research on pharmacokinetic and metabolic profiles of drugs
  • Appearance: solid, off-white to light yellow
Catalog No. 50-004-17649 Supplier Medchemexpress LLC Supplier No. HYN7092S550MG
D-Fructose-4-13C is a 13C labeled form of D-Fructose, a naturally occurring monosaccharide found in many plants. It is a solid with an off-white to light yellow appearance and is suitable for research applications. Stable heavy isotopes, such as those in this product, are used as tracers for quantitation during drug development processes.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Supports research on pharmacokinetic and metabolic profiles of drugs
  • Appearance: solid, off-white to light yellow