Learn More
Medchemexpress LLC β-D-Fructose | 53188-23-1 | 99.8% | 180.16 | 50 MG
SDP
Catalog No. 5000436687
Encompass_Preferred
β-D-Fructose is a β-fruit sugar that can be produced by the hydrolysis of sucrose. It can be used as a sweetness potentiator. β-D-Fructose can also be designed as units of antiproliferative agents against breast (MCF-7) and colon (MDST8) cancer cell lines.
  • Can be produced by the hydrolysis of sucrose.
  • Can be used as a sweetness potentiator.
  • Can be designed as antiproliferative agents.
  • For research use only.
Catalog No. 50-004-36687 Supplier Medchemexpress LLC Supplier No. HY11331950MG
β-D-Fructose is a β-fruit sugar that can be produced by the hydrolysis of sucrose. It can be used as a sweetness potentiator. β-D-Fructose can also be designed as units of antiproliferative agents against breast (MCF-7) and colon (MDST8) cancer cell lines.
  • Can be produced by the hydrolysis of sucrose.
  • Can be used as a sweetness potentiator.
  • Can be designed as antiproliferative agents.
  • For research use only.