Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose | 57-48-7 | 1 G
SDP

Catalog No. 5000420158
Encompass_Preferred

D-Fructose is a monosaccharide found in honey, fruits, and other plants. Two D-Fructose molecules can combine to form the disaccharide sucrose. When ingested orally, it is initially absorbed by the small intestine and then flows through the liver, where its concentration is higher than in blood vessels.

  • Purity of 98.0% or greater
  • Appears as a white to off-white solid
  • Human endogenous metabolite
  • Powder storage conditions: -20°C for 3 years, 4°C for 2 years
  • In solvent storage conditions: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-20158 Supplier Medchemexpress LLC Supplier No. HYN70921G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose is a monosaccharide found in honey, fruits, and other plants. Two D-Fructose molecules can combine to form the disaccharide sucrose. When ingested orally, it is initially absorbed by the small intestine and then flows through the liver, where its concentration is higher than in blood vessels.

  • Purity of 98.0% or greater
  • Appears as a white to off-white solid
  • Human endogenous metabolite
  • Powder storage conditions: -20°C for 3 years, 4°C for 2 years
  • In solvent storage conditions: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.