Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose | 57-48-7 | 1 ML
SDP

Catalog No. 5000418701
Encompass_Preferred

D-Fructose is a monosaccharide found in honey, fruits, and other plants. Two D-Fructose molecules can combine to form a disaccharide, sucrose.

  • Purity of ≥98.0%
  • Molecular weight of 180.16
  • Chemical formula C6H12O6
  • Stable under recommended storage conditions
  • Human endogenous metabolite

Catalog No. 50-004-18701 Supplier Medchemexpress LLC Supplier No. HYN709210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose is a monosaccharide found in honey, fruits, and other plants. Two D-Fructose molecules can combine to form a disaccharide, sucrose.

  • Purity of ≥98.0%
  • Molecular weight of 180.16
  • Chemical formula C6H12O6
  • Stable under recommended storage conditions
  • Human endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.