Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose | 57-48-7 | 10 G
SDP

Catalog No. 5000419892
Encompass_Preferred

D-Fructose is a monosaccharide naturally occurring in honey, fruits, and various plants. It plays a role in forming the disaccharide sucrose when two D-Fructose molecules combine. This product is intended for research use only and is not sold to patients.

  • Molecular weight: 180.16
  • Chemical formula: C6H12O6
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Plants, Gramineae, Saccharum officinarum
  • Endogenous metabolite
  • Shipped at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Biological activity involves absorption by the small intestine and flow through the liver, with higher concentration in the liver.

Catalog No. 50-004-19892 Supplier Medchemexpress LLC Supplier No. HYN709210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose is a monosaccharide naturally occurring in honey, fruits, and various plants. It plays a role in forming the disaccharide sucrose when two D-Fructose molecules combine. This product is intended for research use only and is not sold to patients.

  • Molecular weight: 180.16
  • Chemical formula: C6H12O6
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Plants, Gramineae, Saccharum officinarum
  • Endogenous metabolite
  • Shipped at room temperature in continental US; may vary elsewhere
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Biological activity involves absorption by the small intestine and flow through the liver, with higher concentration in the liver.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.