Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Fructose | 57-48-7 | 5 G
SDP

Catalog No. 5000419678
Encompass_Preferred

D-Fructose is a monosaccharide found naturally in honey, fruits, and other plants. It is initially absorbed by the small intestine and then flows through the liver, resulting in a higher concentration in the liver. It has a molecular weight of 180.16 and a chemical formula of C6H12O6. It appears as a solid, white to off-white in color.

  • Found in honey, fruits, and other plants
  • Monosaccharide
  • Solid, white to off-white appearance
  • Molecular weight of 180.16
  • Chemical formula C6H12O6
  • High purity (98.0%)
  • Endogenous metabolite
  • Can be combined to form sucrose

Catalog No. 50-004-19678 Supplier Medchemexpress LLC Supplier No. HYN70925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Fructose is a monosaccharide found naturally in honey, fruits, and other plants. It is initially absorbed by the small intestine and then flows through the liver, resulting in a higher concentration in the liver. It has a molecular weight of 180.16 and a chemical formula of C6H12O6. It appears as a solid, white to off-white in color.

  • Found in honey, fruits, and other plants
  • Monosaccharide
  • Solid, white to off-white appearance
  • Molecular weight of 180.16
  • Chemical formula C6H12O6
  • High purity (98.0%)
  • Endogenous metabolite
  • Can be combined to form sucrose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.